Learn More
Tetrabenazine, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Specifications
Specifications
| Chemical Name or Material | Tetrabenazine |
| CAS | 58-46-8 |
| Quantity | 250 mg |
| Melting Point | 124°C to 130°C |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C19 H27 N O3 |
| Physical Form | Crystalline Powder |
| Color | White to Light-yellow to Beige |
| Purity | ≥97.5% |