Learn More
STEMCELL Technologies TeSR™-E7™ Medium for Reprogramming (2-Component)
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- TeSR™-E7™ (2-Component) is an animal component-free and defined reprogramming culture medium optimized for the generation of human iPS cells without the use of feeders
- It is based on the E7 formulation published by the laboratory of Dr
- James Thomson (University of Wisconsin-Madison)
Specifications
Specifications
Cell Type | Pluripotent Stem Cells |
For Use With (Application) | Cell Culture, Reprogramming |
Quantity | 500 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies TeSR™-E7™ Medium for Reprogramming (2-Component)