Learn More
Horst™ Temperature Regulating Power Controllers
Brand: Horst™ 041001
This item is not returnable.
View return policy
Specifications
Electronic Triac power control |
HRT005 ELECTRONIC POWER CONTROL
File size: 1.4 MB
Product Content Correction
Product Title
Horst™ Temperature Regulating Power Controllers > Model: HLE 10; continuous adjustment, 0 to 100%, of resistive loads up to 2300w