Learn More
Technical Buffer pH 4.01, Reagecon™ Product Code.: 11424619
Technical Buffer pH 4.01, Reagecon™
BIOPHARMA
Product Code. 11424619 Shop All Reagecon Products
1 L, 1 L
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11424619
Brand: Reagecon TB4011
Specifications
Color Yellow
pH 4.01
Quantity 1 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Product Code
Lot Number4C25C2 Certificate TypeCertificate of Analysis Date29/07/2025 Product CodeTB4011
Lot Number4C24E1 Certificate TypeCertificate of Analysis Date14/12/2024 Product CodeTB4011
Lot Number4C22A1 Certificate TypeCertificate of Analysis Date16/02/2022 Product CodeTB4011
Product Content Correction
Product Title
Technical Buffer pH 4.01, Reagecon™ >