missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TCS 21311
Description
TCS 21311 is a potent JAK3 inhibitor (IC50 = 8 nM). Selective for JAK3 over JAK1, JAK2 and TYK2 (IC50 values are 1017, 2550 and 8055 nM respectively). Also inhibits GSK-3β, PKCα and PKCθ (IC50 values are 3, 13 and 68 nM respectively).
Specifications
Specifications
| Chemical Name or Material | 3-[5-[4-(2-Hydroxy-2-methyl-1-oxopropyl)-1-piperazinyl]-2-(trifluoromethyl)phenyl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione |
| CAS | 1260181-14-3 |
| Quantity | 10 mg |
| Target | JAK Kinase Inhibitors |
| Molecular Formula | C27H25F3N4O4 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?