Learn More
TBE Sample Buffer, 6X, Thermo Scientific™ Product Code.: 15493139
TBE Sample Buffer, 6X, Thermo Scientific™
Product Code. 15493139
25 mL, 25mL
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
25mL
50mL
Product Code. 15493139
Brand: Thermo Scientific Alfa Aesar J60357.AC
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sample Loading Buffer
Color Blue
Concentration 6X
Physical Form Liquid
Quantity 25 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ30L030 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeJ60357.AC,J60357.AD
Lot NumberP23I023 Certificate TypeCertificate of Analysis Date24/02/2023 Product CodeJ60357.AC,J60357.AD
Lot NumberW01I012 Certificate TypeCertificate of Analysis Date19/01/2023 Product CodeJ60357.AC,J60357.AD
Lot NumberP17I057 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeJ60357.AC,J60357.AD
Lot NumberQ02E503 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeJ60357.AC,J60357.AD
Product Content Correction
Product Title
TBE Sample Buffer, 6X, Thermo Scientific™ >