Learn More
Tocris Bioscience™ TASP 0277308
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
TASP 0277308 is a potent and selective sphingosine 1-phosphate 1 (S1P1) receptor antagonist (IC50 = 7.8 nM). TASP 0277308 inhibits S1P-induced chemotaxis (IC50 = 1.1-1.6 nM), HUVEC cell proliferation (IC50 = 3-3.6 nM) and suppresses angiogenesis in vivo. TASP 0277308 also blocks VEGF-induced tube-formation of HUVECs in vitro and reverses or impairs the development of arthritis or mechano-allodynia.
Specifications
Specifications
| Chemical Name or Material | 3,4-Dichloro-N-[(1 R)-1-[4-ethyl-5-[3-(4-methyl-1-piperazinyl)phenoxy]-4 H-1,2,4-triazol-3-yl]ethyl]benzenesulfonamide |
| CAS | 945725-50-8 |
| Quantity | 5 mg |
| Target | Sphingosine-1-phosphate Receptor Antagonists |
| Molecular Formula | C23H28Cl2N6O3S |
| Purity | 0.98 |