Learn More
Applied Biosystems™ TaqMan™ GMO Maize 35S Detection Kit, with protocol Product Code.: 12079800
Applied Biosystems™ TaqMan™ GMO Maize 35S Detection Kit, with protocol
Product Code. p-7180624
1 Kit, Each
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. 12079800
Brand: Applied Biosystems™ 4327690
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Kit contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods, and their ingredients.
  • Contains reagents needed to reliably detect the presence of genetically modified organism (GMO)-specific DNA sequences in seed, grain, processed foods, and their ingredients
  • Universal assay detects and quantifies all approved genetically modified organisms (GMOs) in Europe, and the vast majority of GMOs approved in other countries
  • Fluorogenic 5' nuclease assay using TaqMan™ probes provides accurate and reproducible quantitative results
  • Comprehensive controls provide high confidence in results
  • Innovative GMO quantitation software allows users to quickly and easily determine the percentage of GMO content
  • Offers the complete solution including sample preparation, instruments, reagents, analysis software, technical support, instrument service, and GMO expertise
TRUSTED_SUSTAINABILITY
Specifications
Detection Method Primer-Probe
For Use With (Equipment) 7300 System, 7500 System, AB 7900 HT 384-Well System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
Content And Storage Each kit contains TaqMan GMO Mix, AmpliTaq Gold™ DNA Polymerase, positive and negative controls. Each kit contains enough regents for 100 assays.
Store all components at -25 to -15°C.
Shipping Condition Dry Ice
Sample Type Food, Seed
Target Organism Class Maize, Genetically Modified
Quantity 1 Kit
Product Line TaqMan™
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2201097 Certificate TypeCertificate of Analysis Date14/02/2022 Product Code4327690
Lot Number2112096 Certificate TypeCertificate of Analysis Date17/12/2021 Product Code4327690
Lot Number2106095 Certificate TypeCertificate of Analysis Date17/06/2021 Product Code4327690
Lot Number1902092 Certificate TypeCertificate of Analysis Date11/03/2019 Product Code4327690
Lot Number1901091 Certificate TypeCertificate of Analysis Date15/01/2019 Product Code4327690
Product Content Correction
Product Title
Applied Biosystems™ TaqMan™ GMO Maize 35S Detection Kit, with protocol >