missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies SU5402

Product Code. p-7217503
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 17130292

Brand: STEMCELL Technologies 73912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MEK/ERK pathway inhibitor; Inhibits VEGFR2, FGFR1, and PDGFRB

  • SU5402 is a potent and selective inhibitor of vascular endothelial growth factor receptor 2 (VEGFR-2), fibroblast growth factor receptor 1 (FGFR-1), and platelet-derived growth factor receptor beta (PDGFRB) with IC50 = 0.02, 0.03, and 0.51 μM, respectively (Sun et al.)

Specifications

CAS 215543-92-3
Quantity 1 mg
Molecular Formula C17H16N2O3
Purity ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.