missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 710 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
986.00 NOK - 2050.00 NOK
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | eFluor 710 |
Form | Liquid |
Product Line | eBioscience™ |
Product Type | Streptavidin Conjugate (Fluorescent) |
Alert:
Contains Azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15331840
|
Invitrogen™
49-4317-80 |
25 μg |
986.00 NOK
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15341840
|
Invitrogen™
49-4317-82 |
100 μg |
2050.00 NOK
100µg |
In Stock
Log in to see stock. |
|||||
Specifications
0.2 mg/mL | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
eFluor 710 |
eFluor 710 | |
eBioscience™ | |
Biotin |
For Research Use Only