Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 710 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 49-4317-82
Additional Details : Weight : 0.25000kg
Alert:
Contains Azide
Specifications
0.2 mg/mL | |
100 μg | |
eBioscience™ | |
Biotin |
eFluor 710 | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
eFluor 710 |
For Research Use Only