Learn More
Tocris Bioscience™ STF 31
Product Code. 18024552
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18024552 10 mg 10mg
18095532 50 mg 50mg
2 options
Product Code. 18024552
Brand: Tocris Bioscience™ 4484/10
NAMPT inhibitor; also GLUT1 inhibitor
Inhibitor of GLUT1; inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells. Activity causes necrotic cell death in von Hippel-Lindau (VHL)-deficient RCC cells. Also NAMPT inhibitor. Eliminates human pluripotent stem cells from culture with limited toxicity towards differentiated cells.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 724741-75-7
Molecular Formula C23H25N3O3S
Specifications
Quantity 10 mg
Formula Weight 423.53
Purity >99%
Molecular Weight (g/mol) 423.531
Chemical Name or Material STF 31