Learn More
Tocris Bioscience™ SR 13800
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SR 13800 is a potent MCT1 inhibitor (IC50 = 0.5 nM); blocks proliferation of Raji lymphoma cells in vitro. Also inhibits lactate uptake in breast cancer cells in vitro.
Specifications
Specifications
| Chemical Name or Material | 2,6-Dihydro-7-[(3-hydroxypropyl)thio]-2-methyl-4-(2-methylpropyl)-6-(1-naphthalenylmethyl)-1H-pyrrolo[3,4-d]pyridazin-1one |
| CAS | 227321-12-2 |
| Quantity | 50 mg |
| Target | Monocarboxylate Transporter Inhibitors |
| Molecular Formula | C25H29N3O2S |
| Formula Weight | 435.58 |
| Purity | >98% |