Learn More
Fisherbrand™ Sound Protection Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Shop All Fisherbrand ProductsSpecifications
Specifications
| Type | Sound Protection Cover |
| For Use With (Equipment) | Ultrasonic Cleaner, Elmasonic Xtra ST 2500 H |
| Style | Hinged Lid |
| For Use With (Application) | Noise reduction for ultrasonic cleaning units |
| Material | Stainless Steel |