Learn More
Memmert™ Software Atmocontrol, Base License For 1 Unit, Iq/Oq Doc. Available In De And En, For Hcp50
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand:  Memmert™ HCP50FDAQ1
29937.18 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16394877
  • 32935.00 NOK / Each
Only null left
Estimated Shipment: 28-10-2025
Description
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base License for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106
Software
Base License for 1 Unit
Videos
  • Memmert AtmoCONTROL (EN)
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Memmert AtmoCONTROL (EN)
    • Memmert ControlCOCKPIT (EN)
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Memmert ControlCOCKPIT (EN)
      Certificates
      Promotions
      Promotions
      Product Content Correction
      Product Title
      Memmert™ Software Atmocontrol, Base License For 1 Unit, Iq/Oq Doc. Available In De And En, For Hcp50 >