missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hexanitrocobaltate(III), ACS Reagent, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Melting Point | 220°C |
Color | Golden or Orange-Yellow |
Quantity | 100 g |
Linear Formula | Na3Co(NO2)6 |
UN Number | UN1479 |
Formula Weight | 403.94g/mol |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Appearance | Powder |
Chemical Name or Material | Sodium Hexanitrocobaltate(III) |
Safety and Handling



Product Identifier
- Sodium hexanitrocobaltate(III)
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Trisodium hexanitritocobaltate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hexanitrocobaltate(III), ACS Reagent, Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction