Learn More
STEMCELL Technologies SIS3
Product Code. p-7217814
Encompass_Preferred
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
Product Code. 17120302
Brand: STEMCELL Technologies 73962
Activin/NODAL/TGF-β pathway inhibitor; Inhibits SMAD3
  • SIS3 (specific inhibitor of SMAD3) is a cell-permeable selective inhibitor of transforming growth factor β1 (TGF-β1)-dependent SMAD3 phosphorylation (IC50 = 3 μM) and SMAD3-mediated cellular signaling (Jinnin et al.) SIS3 has no effect on SMAD2, p38 mitogen-activated protein kinase (MAPK), extracellular signal-regulated kinase (ERK), or phosphoinositide 3-kinases
  • Facilitates the production of anterior ectoderm from mouse induced pluripotent stem cells (Oshima et al.)
  • Inhibits myofibroblast differentiation of normal human dermal fibroblasts (Jinnin et al.)
Specifications
CAS 1009104-85-1
Synonym Smad3 Inhibitor
Quantity 500 μg
Molecular Formula C28H27N3O3
Purity ≥98%
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.