Learn More
Vitlab™ Simplex II Bottletop Dispenser
Vitlab™ Simplex II Bottletop Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
3683.95 NOK - 7130.64 NOK
Products 5
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15670915
View Documents
Vitlab™
1621503
0.2 to 2.0 mL
ONLINE EXCLUSIVE PRICE
4010.00 NOK
3683.95 NOK / Each
Save 326.05 NOK
8% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 23-12-2025
Add to basket
15624854
View Documents
Vitlab™
1621504
0.5 to 5.0 mL
ONLINE EXCLUSIVE PRICE
4010.00 NOK
3683.95 NOK / Each
Save 326.05 NOK
8% Off
View alternative products
Save Up to 
»
15634854
View Documents
Vitlab™
1621505
1.0 to 10.0 mL
ONLINE EXCLUSIVE PRICE
4010.00 NOK
3683.95 NOK / Each
Save 326.05 NOK
8% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 30-10-2025
Add to basket
15680915
View Documents
Vitlab™
1621506
2.5 to 25 mL
ONLINE EXCLUSIVE PRICE
5245.00 NOK
4817.67 NOK / Each
Save 427.33 NOK
8% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 28-10-2025
Add to basket
15600925
View Documents
Vitlab™
1621508
10.0 to 100.0 mL
ONLINE EXCLUSIVE PRICE
7760.00 NOK
7130.64 NOK / Each
Save 629.36 NOK
8% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 28-10-2025
Add to basket
Description
Description
  • Direct displacement plunger with broad PFA seal prevents crystallisation build-up
  • Settings are quickly and accurately adjusted due to interior scalloped track and practical screw mechanism
  • Screwable ventilation plug for easy connection of a drying tube
  • Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
  • Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
  • Nominal volume 2 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
  • Nominal volume 25 to 100mL includes adapters GL 32, GL 38, S 40 and telescopic intake tube (length 170 to 330mm)
Documents
Product Certifications
Product Content Correction
Product Title
Vitlab™ Simplex II Bottletop Dispenser