missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC10 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
2060.00 NOK - 4510.00 NOK
Specifications
| Antigen | SIGLEC10 |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17614036
|
Invitrogen™
MA543575 |
20 μL |
2060.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17604036
![]() |
Invitrogen™
MA543574 |
100 μL |
4510.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.Specifications
| SIGLEC10 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940 | |
| SIGLEC10 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96LC7 | |
| 89790 | |
| Recombinant SIGLEC10 (Met571-Gln697) expressed in E.coli. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SIGLEC10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title