missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC10 Monoclonal Antibody (C5), Invitrogen™

Description
SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
Specifications
Specifications
| Antigen | SIGLEC10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | C5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | SIGLEC10 |
| Gene Accession No. | Q96LC7 |
| Gene Alias | PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?