missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SIGLEC10 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543575

 View more versions of this product

Product Code. 17614036

  • 2060.00 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SIGLECs are members of the immunoglobulin superfamily that are expressed on the cell surface. Most SIGLECs have 1 or more cytoplasmic immune receptor tyrosine-based inhibitory motifs, or ITIMs. SIGLECs are typically expressed on cells of the innate immune system, with the exception of the B-cell expressed SIGLEC6.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIGLEC10
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q96LC7
SIGLEC10
Recombinant SIGLEC10 (Met571-Gln697) expressed in E.coli.
20 μL
Primary
Human
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Western Blot
C5
Unconjugated
SIGLEC10
PRO940; sialic acid binding Ig like lectin 10; sialic acid binding Ig-like lectin 10; sialic acid binding Ig-like lectin 10 Ig-like lectin 7; sialic acid binding Ig-like lectin G; sialic acid-binding Ig-like lectin 10; Siglec; SIGLEC10; Siglec-10; Siglecg; siglec-like gene 2; siglec-like protein 2; SLG2; UNQ477/PRO940
Mouse
Protein A/G
RUO
89790
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.