Learn More
Tocris Bioscience™ SET 2
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SET 2 is a TRPV2 (transient receptor potential vanilloid type 2) antagonist (IC50 = 0.46 μM). Selective for TRPV2 over TRPV1, TRPV3 and TRPV4 expressed in HEK293 cells. Inhibits lyosphosphatidic acid (LPA)-induced prostate cancer cell migration in vitro.
Specifications
Specifications
| Chemical Name or Material | N-(2-Furanylmethyl)-3-[[4-(methylpropylamino)-6-(trifluoromethyl)-2-pyrimidinyl]thio]propanamide |
| CAS | 2313525-20-9 |
| Quantity | 50 mg |
| Target | TRPV Antagonists |
| Molecular Formula | C17H21F3N4O2S |
| Purity | 0.98 |