missing translation for 'onlineSavingsMsg'
Learn More

Melford Separating or Resolving Buffer 4X Solution

4X concentrated buffer solution for preparation of Laemmli SDS-PAGE resolving gels.

Brand:  Melford S28100-500.0

764.75 NOK valid until 2025-08-31
Use promo code "24128" to get your promotional price.


Product Code. 30018812

  • 805.00 NOK / 500mL
Science Is Hard Enough Your Bioreagents Shouldn’t Be
Use promo code 24128 at checkout
View Available Promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

4X concentrated buffer solution for preparation of Laemmli SDS-PAGE resolving gels.

4X concentrated buffer solution for preparation of Laemmli SDS-PAGE resolving gels.
Specifications

Specifications

7732-18-5
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Melford Separating or Resolving Buffer 4X Solution > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.