Learn More
3M™ Secure Click™ Reusable Full Face Respirator Product Code.: 17966640
Only undefined left
Add to basket
3M™ Secure Click™ Reusable Full Face Respirator
Product Code. 17966640
Large, Each
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
Product Code. 17966640
Brand: 3M™ 7100262626
Only undefined left
Add to basket
Only undefined left
Add to basket
The Secure Click™ Reusable Full Face Mask FF-800 Series helps protect against particulates and a wide variety of gases and vapors when used with approved Secure Click™ filters.
  • The quick, easy, one-touch positive pressure push button seal check helps provide improved confidence in respirator fit
  • Two dual-flow gas and vapor filters on each respirator combine to create the world's first quad flow gas and vapor filter system with four airflow paths providing easy breathing and comfort
  • Large lens for excellent field of vision, and treated with Scotchgard™ Protector for improved paint and stain resistance
  • Six-strap harness helps provide a secure comfortable fit
  • Speaking diaphragm is designed to help provide easy communication
  • Recommended applications include assembly, chemical clean-up, chemical handling, chipping, chiseling, cleaning, furnace operations, grinding, machining, masonry, painting, pouring/casting, sanding and sawing.
  • Available in three sizes
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Filter Type Secure Click™ Filters
Size Large
Product Content Correction
Product Title
3M™ Secure Click™ Reusable Full Face Respirator >