Learn More
Tocris Bioscience™ SD 208
GREENER_CHOICE
Product Code. 18065912
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18065912
Brand: Tocris Bioscience™ 3269/10
Potent ATP-competitive TGF-βRI inhibitor
Potent, orally active ATP-competitive transforming growth factor-β receptor 1 (TGF-βRI) inhibitor (IC50 = 49 nM). Displays > 100-fold and > 17-fold selectivity over TGF-βRII and other common kinases respectively. Exhibits anti-inflammatory and antitumor activity. Also promotes an antiglioma immune response. Ameliorates germinal matrix hemorrhage-induced neurological deficits in neonatal rats.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 627536-09-8
Molecular Formula C17H10ClFN6
Specifications
Quantity 10 mg
Formula Weight 352.75
Purity >98%
Molecular Weight (g/mol) 352.76
Chemical Name or Material SD 208