missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCH 336
Description
SCH 336 is a highly potent and selective CB2 receptor inverse agonist (Ki = 1.8 nM, EC50 = 2 nM). Displays 100-fold selectivity for CB2 receptors over CB1. Increases forskolin stimulated cAMP accumulation in CHO cells expressing human CB2 receptors. Inhibits leukocyte migration in a murine model of delayed-type hypersensitivity and inhibits antigen-induced lung eosinophilia in a mouse allergy model.
Specifications
Specifications
| Chemical Name or Material | N-[(1S)-1-[4-[[4-Methoxy-2-[(4-methoxyphenyl)sulfonyl]phenyl]sulfonyl]phenyl]ethyl]methanesulfonamide |
| CAS | 447459-51-0 |
| Assay Percent Range | >98% |
| Quantity | 10 mg |
| Molecular Formula | C23H25NO8S3 |
| Formula Weight | Observed MW: 539.64 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?