Learn More
Tocris Bioscience™ SCH 336
Product Code. 18783998
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18783998 10 mg 10mg
18720148 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18783998 Supplier Tocris Bioscience™ Supplier No. 5815/10
Highly potent and selective CB2 inverse agonist
SCH 336 is a highly potent and selective CB2 receptor inverse agonist (Ki = 1.8 nM, EC50 = 2 nM). Displays 100-fold selectivity for CB2 receptors over CB1. Increases forskolin stimulated cAMP accumulation in CHO cells expressing human CB2 receptors. Inhibits leukocyte migration in a murine model of delayed-type hypersensitivity and inhibits antigen-induced lung eosinophilia in a mouse allergy model.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[(1S)-1-[4-[[4-Methoxy-2-[(4-methoxyphenyl)sulfonyl]phenyl]sulfonyl]phenyl]ethyl]methanesulfonamide
CAS 447459-51-0
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C23H25NO8S3
Formula Weight Observed MW: 539.64