missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 265610
GREENER_CHOICE

Product Code. 18044432
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18044432 10 mg 10mg
30001412 1 mg 1mg
18094512 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18044432 Supplier Tocris Bioscience™ Supplier No. 2724/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent CXCR2 antagonist

Potent CXCR2 antagonist that inhibits CINC-1-mediated but not C5a-mediated Ca2+ mobilization (IC50 values are 3.4 and 6800 nM respectively). Inhibits CINC-induced chemotaxis and attenuates neutrophil accumulation in inflammatory lung injury in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 211096-49-0
Molecular Formula C14H9BrN6O

Specifications

Quantity 10 mg
Formula Weight 357.16
Purity >98%
Molecular Weight (g/mol) 357.171
Chemical Name or Material SB 265610
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.