Learn More
STEMCELL Technologies Rolipram
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
- Rolipram is a cell-permeable, selective inhibitor of Type 4 cyclic nucleotide phosphodiesterases (PDE4), which mediate cyclic AMP (cAMP) degradation
- Rolipram preferably inhibits PDE4 isoform A (IC50 = 3 nM) over other isoforms such as B and D (IC50 = 130 and 240 nM respectively; MacKenzie & Houslay)
- It inhibits interferon (IFN)-γ stimulated phosphorylation of p38 mitogen-activated protein (MAP) kinase through PDE4B and/or PDE4D isoform inhibition (MacKenzie & Houslay)
Specifications
Specifications
| CAS | 61413-54-5 |
| Synonym | SB 95952; ZK 62711 |
| Quantity | 25 mg |
| Molecular Formula | C16H21NO3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.