missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Remodelin
GREENER_CHOICE

Product Code. p-7110788
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18762459

Brand: Tocris Bioscience™ 5193/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-acetyltransferase 10 (NAT10) inhibitor

Remodelin is a N-acetyltransferase 10 (NAT10) inhibitor. Improves nuclear shape in HGPS (Hutchinson-Gilford progeria syndrome) cells and lamin A/C depleted cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-(2-Cyclopentylidenehydrazinyl)-4-thiazolyl]benzonitrile
CAS 949912-58-7
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C15H14N4S
Formula Weight 282.36
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.