Learn More
Refractive Index 1.420087 Equivalent, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Specifications
Specifications
| Packaging | 15mL |
| Quantity | 15 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
| Lot Number | Certificate Type | Date | Product Code |
|---|---|---|---|
| RIBS2025E1 | Certificate of Analysis | 16/09/2025 | RIBS20S |
| RIBS2024J1 | Certificate of Analysis | 01/12/2024 | RIBS20S |
| RIBS2024F1 | Certificate of Analysis | 19/06/2024 | RIBS20S |
| RIBS2024C1 | Certificate of Analysis | 23/05/2024 | RIBS20S |
| RIBS2023L1 | Certificate of Analysis | 12/12/2023 | RIBS20S |
Product Content Correction
Product Title
Refractive Index 1.420087 Equivalent, Reagecon™ >