Learn More
enQuireBio™ Recombinant Human Mitochondrial cardiolipin hydrolase Protein
A DNA sequence encoding the Homo sapiens (Human) Mitochondrial cardiolipin hydrolase, was expressed in the hosts and tags indicated.
2526.25 NOK - 34365.00 NOK
Specifications
Accession Number | NP_849158.2 |
---|---|
Gene ID (Entrez) | 201164 |
Name | Mitochondrial cardiolipin hydrolase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091341
|
enQuireBio™
QP7879-EC-100UG |
100 μg |
7745.00 NOK
100µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011351
|
enQuireBio™
QP7879-EC-1MG |
1 mg |
34365.00 NOK
1mg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001351
|
enQuireBio™
QP7879-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021351
|
enQuireBio™
QP7879-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031351
|
enQuireBio™
QP7879-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041351
|
enQuireBio™
QP7879-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_849158.2 | |
Mitochondrial cardiolipin hydrolase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
201164 | |
Research Use Only | |
PLD6 | |
Recombinant Protein | |
E. coli | |
MGRLSWQVAA AAAVGLALTL EALPWVLRWL RSRRRRPRRE ALFFPSQVTC TEALLRAPGA ELAELPEGCP CGLPHGESAL SRLLRALLAA RASLDLCLFA FSSPQLGRAV QLLHQRGVRV RVVTDCDYMA LNGSQIGLLR KAGIQVRHDQ DPGYMHHKFA IVDKRVLITG SLNWTTQAIQ NNRENVLITE DDEYVRLFLE EFERIWEQFN PTKYTFFPPK KSHGSCAPPV SRAGGRLLSW HRTCGTSSES QT | |
Greater than 90% as determined by SDS-PAGE. |