missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mitochondrial cardiolipin hydrolase Protein
A DNA sequence encoding the Homo sapiens (Human) Mitochondrial cardiolipin hydrolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7879-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_849158.2 | |
Mitochondrial cardiolipin hydrolase Protein | |
Research Use Only | |
PLD6 | |
Recombinant Protein | |
E. coli | |
MGRLSWQVAA AAAVGLALTL EALPWVLRWL RSRRRRPRRE ALFFPSQVTC TEALLRAPGA ELAELPEGCP CGLPHGESAL SRLLRALLAA RASLDLCLFA FSSPQLGRAV QLLHQRGVRV RVVTDCDYMA LNGSQIGLLR KAGIQVRHDQ DPGYMHHKFA IVDKRVLITG SLNWTTQAIQ NNRENVLITE DDEYVRLFLE EFERIWEQFN PTKYTFFPPK KSHGSCAPPV SRAGGRLLSW HRTCGTSSES QT | |
Greater than 90% as determined by SDS-PAGE. |
201164 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |