Learn More
enQuireBio™ Recombinant Human Mitochondrial cardiolipin hydrolase Protein
A DNA sequence encoding the Homo sapiens (Human) Mitochondrial cardiolipin hydrolase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7879-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16011351
  • 34365.00 NOK / 1mg
Estimated Shipment: 09-07-2024
Specifications
NP_849158.2
Mitochondrial cardiolipin hydrolase Protein
Research Use Only
PLD6
Recombinant Protein
E. coli
MGRLSWQVAA AAAVGLALTL EALPWVLRWL RSRRRRPRRE ALFFPSQVTC TEALLRAPGA ELAELPEGCP CGLPHGESAL SRLLRALLAA RASLDLCLFA FSSPQLGRAV QLLHQRGVRV RVVTDCDYMA LNGSQIGLLR KAGIQVRHDQ DPGYMHHKFA IVDKRVLITG SLNWTTQAIQ NNRENVLITE DDEYVRLFLE EFERIWEQFN PTKYTFFPPK KSHGSCAPPV SRAGGRLLSW HRTCGTSSES QT
Greater than 90% as determined by SDS-PAGE.
201164
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol