Learn More
enQuireBio™ Recombinant Human IL7 / Interleukin 7 Protein
Recombinant Protein
2096.25 NOK - 74865.00 NOK
Specifications
Accession Number | NP_000871.1 |
---|---|
Gene ID (Entrez) | 3574 |
Name | IL7 / Interleukin 7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948418
|
enQuireBio™
QP1180-500UG |
500 μg |
45570.00 NOK
500µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938418
|
enQuireBio™
QP1180-1MG |
1 mg |
74865.00 NOK
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918418
|
enQuireBio™
QP1180-10 UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15928418
|
enQuireBio™
QP1180-100 UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000871.1 | |
IL7 / Interleukin 7 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the Mouse 2E8 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg. | |
Human | |
Untagged | |
10 mM Aceitic acid |
3574 | |
Research Use Only | |
IL7 | |
Recombinant Protein | |
E. coli | |
MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH | |
Greater than 95% as determined by SDS-PAGE. |