missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL7 / Interleukin 7 Protein
Recombinant Protein
Brand: enQuireBio™ QP1180-10 ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000871.1 | |
IL7 / Interleukin 7 Protein | |
Research Use Only | |
IL7 | |
Recombinant Protein | |
E. coli | |
MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH | |
Greater than 95% as determined by SDS-PAGE. |
3574 | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 1 ng/mL in the Mouse 2E8 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg. | |
Human | |
Untagged | |
10 mM Aceitic acid |