Learn More
enQuireBio™ Recombinant H. pylori Orotidine 5'-phosphate decarboxylase Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Orotidine 5'-phosphate decarboxylase, was expressed in the hosts and tags indicated. option, abo
3192.75 NOK - 34365.00 NOK
Specifications
Accession Number | NP_206807.1 |
---|---|
Gene ID (Entrez) | 898802 |
Name | Orotidine 5'-phosphate decarboxylase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086491
|
enQuireBio™
QP8793-EC-100UG |
100 μg |
9910.00 NOK
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006501
|
enQuireBio™
QP8793-EC-1MG |
1 mg |
34365.00 NOK
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096491
|
enQuireBio™
QP8793-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016501
|
enQuireBio™
QP8793-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026501
|
enQuireBio™
QP8793-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036501
|
enQuireBio™
QP8793-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_206807.1 | |
Orotidine 5'-phosphate decarboxylase Protein | |
Not determined. | |
Full Length | |
H. pylori | |
His | |
Tris-based buffer, 50% glycerol |
898802 | |
Research Use Only | |
pyrF | |
Recombinant Protein | |
E. coli | |
MQLCVALDLE KKEDNLSLLQ ELKGLDLWAK VGLRSFIRDG AVFLDEIRKI DENFKIFLDL KLYDIPYTMA NAALECAKLD IDMLTVHLSS AKSALTALMQ RLNALKKRPL IMGVSALTSF SEEEFLMVYN APLKTQAIKL SAMGKESGID GVVCSVFESL AIKEALGKDF LTLTPGIRLD QNDKEDQERV ANAKEAKQNL SDFIVVGRPI YQAKEPREVV LELLKDC | |
Greater than 90% as determined by SDS-PAGE. |