Learn More
enQuireBio™ Recombinant H. pylori Orotidine 5'-phosphate decarboxylase Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Orotidine 5'-phosphate decarboxylase, was expressed in the hosts and tags indicated. option, abo
Brand: enQuireBio™ QP8793-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_206807.1 | |
Orotidine 5'-phosphate decarboxylase Protein | |
Research Use Only | |
pyrF | |
Recombinant Protein | |
E. coli | |
MQLCVALDLE KKEDNLSLLQ ELKGLDLWAK VGLRSFIRDG AVFLDEIRKI DENFKIFLDL KLYDIPYTMA NAALECAKLD IDMLTVHLSS AKSALTALMQ RLNALKKRPL IMGVSALTSF SEEEFLMVYN APLKTQAIKL SAMGKESGID GVVCSVFESL AIKEALGKDF LTLTPGIRLD QNDKEDQERV ANAKEAKQNL SDFIVVGRPI YQAKEPREVV LELLKDC | |
Greater than 90% as determined by SDS-PAGE. |
898802 | |
10 μg | |
Not determined. | |
Full Length | |
H. pylori | |
His | |
Tris-based buffer, 50% glycerol |