missing translation for 'onlineSavingsMsg'
Learn More
Learn More
exendin-4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of exendin-4.
Brand: Abnova PAB5186.150ug
This item is not returnable.
View return policy
Description
Sequence: HGEGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPSSpecifications
| exendin-4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of exendin-4. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| A synthetic peptide (conjugated with KLH) corresponding to exendin-4. | |
| RUO | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
exendin-4, Rabbit, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction