Learn More
Tocris Bioscience™ PFK 15
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PFK 15 is an originally reported to be a selective PFKFB3 inhibitor (IC50 = 207 nM); a more recent report failed to demonstrate activity in a PFKFB3 kinase assay. Selective for PFKFB3 over a panel of 96 kinases. Reduces glucose uptake and induces apoptosis in H522 lung adenocarcinoma and Jurkat T cell leukemia cells. Suppresses glucose uptake and tumor growth, and inhibits metastasis of Lewis lung carcinoma cell xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | PFK 15 |
| CAS | 4382-63-2 |
| Synonym | 1-(4-Pyridinyl)-3-(2-quinolinyl)-2-propen-1-one |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C17H12N2O |
| Formula Weight | 260.29 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >99% |
Research Use Only. Not For Human or Veterinary Use