Learn More
Tocris Bioscience™ ML 179
Description
ML 179 is a selective liver receptor homolog 1 (LRH1, NR5A2) inverse agonist (IC50 = 320 nM and maximum efficacy of 64% repression). Exhibits >31-fold selectivity over SF1.
Specifications
Specifications
| Chemical Name or Material | ML 179 |
| CAS | 1883548-87-5 |
| Synonym | 3-Cyclohexyl-6-[4-[3-(trifluoromethyl)phenyl]piperazin-1-yl]-1H-pyrimidine-2,4-dione |
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in DMSO |
| Molecular Formula | C21H25F3N4O2 |
| Formula Weight | 422.44 |
| Physical Form | Crystalline Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >99% |
Research Use Only. Not For Human or Veterinary Use