Learn More
Tocris Bioscience™ Blasticidin S HCl
Description
Blasticidin S HCl is an antibiotic. Selection reagent for bis, bsr and BSD transformed cells.
Specifications
Specifications
| Chemical Name or Material | Blasticidin S HCl |
| CAS | 3-9-3513 |
| Synonym | (S)-4-[[3-Amino-5-[(aminoiminomethyl)methylamino]-1-oxopentyl]amino]-1-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,2,3,4-tetradeoxy-β-D-erythro-hex-2-enopyranuronic acid hydrochloride |
| Quantity | 25 mg |
| Solubility | Soluble to 10mM in water |
| Molecular Formula | C17H26N8O5·HCl |
| Formula Weight | 458.9 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use