Learn More
Tocris Bioscience™ R-96544 hydrochloride
Product Code. 18054682
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18054682
Brand: Tocris Bioscience™ 1742/10
Potent, selective 5-HT2A antagonist
Potent, selective 5-HT2 receptor antagonist; displays some selectivity for 5-HT2A receptors (Ki = 1.6 nM). IC50 values are 2.2, 310, 2400, 3700, > 5000 and > 5000 nM for 5-HT2, α1-adrenergic, D2 dopamine, 5-HT1, 5-HT3 and β-adrenergic receptors respectively. Inhibits 5-HT-induced platelet aggregation and pressor responses in vivo.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 167144-80-1
Molecular Formula C22H30ClNO3
Specifications
Quantity 10 mg
Formula Weight 391.94
Purity >99%
Molecular Weight (g/mol) 391.936
Chemical Name or Material R-96544 hydrochloride