Learn More
Tocris Bioscience™ Pyr 10
Description
Pyr 10 is a TRPC3 (transient receptor potential canonical 3) channel inhibitor (IC50 = 0.72 μM for TRPC3 mediated calcium entry in vitro), which displays approximately 18-fold selectivity for TRPC3-mediated receptor operated calcium entry (ROCE) over STIM1/Orai1-mediated store operated calcium entry (SOCE). Pyr 10 inhibits NFATc3 activation and inhibits proliferation of rat ventricular cardiac fibroblasts.
Specifications
Specifications
| Chemical Name or Material | N-[4-[3,5-bis(Trifluoromethyl)-1 H-pyrazol-1-yl]phenyl]-4-methylbenzenesulfonamide |
| CAS | 1315323-00-2 |
| Quantity | 50 mg |
| Target | TRPC Channel Blockers |
| Molecular Formula | C18H13F6N3O2S |
| Purity | 0.98 |