missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein A Mouse anti-S. aureus, Unconjugated, Clone: 1G6A6, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Protein A |
|---|---|
| Clone | 1G6A6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809265
|
Proteintech
66945-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899255
|
Proteintech
66945-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Protein A antibody from Proteintech is a mouse monoclonal antibody generated with peptide of Protein A. This antibody recognizes staphylococcus aureus antigen. The Protein A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Protein A | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| IgG2a |
| 1G6A6 | |
| Western Blot, Immunoprecipitation, ELISA | |
| Unconjugated | |
| S. aureus | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title