missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein A Mouse anti-S. aureus, Unconjugated, Clone: 1G6A6, Proteintech
Shop All Proteintech ProductsDescription
The Protein A antibody from Proteintech is a mouse monoclonal antibody generated with peptide of Protein A. This antibody recognizes staphylococcus aureus antigen. The Protein A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | Protein A |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Clone | 1G6A6 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Target Species | S. aureus |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?