Learn More
Protein A Mouse anti-S. aureus, Unconjugated, Clone: 1G6A6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66945-1-IG-20UL
This item is not returnable.
View return policy
Description
The Protein A antibody from Proteintech is a mouse monoclonal antibody generated with peptide of Protein A. This antibody recognizes staphylococcus aureus antigen. The Protein A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Protein A | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| IgG2a |
| Western Blot, Immunoprecipitation, ELISA | |
| 1G6A6 | |
| Unconjugated | |
| 20 μL | |
| S. aureus |