Learn More
Sartorius Proline Plus Mechanical Pipette Multipacks Product Code.: 15663094
In Stock
  • ONLINE EXCLUSIVE PRICE
    12880.00 NOK
    10762.88 NOK / 1 set
    Save 2117.12 NOK
    16% Off
Only null left
Add to basket
Sartorius Proline Plus Mechanical Pipette Multipacks
BIOPHARMAGREENER_CHOICE
Product Code. 15663094 Shop All Sartorius Products
Mechanical Pipette Multipacks, 10/Pk., 0.5 to 10 ÎĽL, 2 to 20 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL, 1 set
Click to view available options
Type:
Mechanical Pipette Multipacks
Quantity:
1/Pk.
10/Pk.
20/Pk.
Volume (Metric):
0.5 to 10 ÎĽL, 10 to 100 ÎĽL, 100 to 1000 ÎĽL
0.5 to 10 ÎĽL, 10 to 100 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL
0.5 to 10 ÎĽL, 10 to 100 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL, 500 to 5,000 ÎĽL
0.5 to 10 ÎĽL, 2 to 20 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL
0.5 to 10 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL, 30 to 300 ÎĽL
100 to 1000 ÎĽL, 500 to 5000 ÎĽL, 1 to 10 mL
2 to 20 ÎĽL, 10 to 100 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL, 500 to 5000 ÎĽL
2 to 20 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL
Unit Size:
1 set
Product Code. 15663094
Brand: Sartorius LH728672
  • ONLINE EXCLUSIVE PRICE
    12880.00 NOK
    10762.88 NOK / 1 set
    Save 2117.12 NOK
    16% Off
Only null left
In Stock
Add to basket
Only null left
Add to basket
Includes
4 x Single-channel pipets (0.5 to 10ÎĽL, 2 to 20ÎĽL, 20 to 200ÎĽL, 100 to 1000ÎĽL), Tip Racks of 96 tips (0.1 to 10ÎĽL, 0.5 to 200ÎĽL, 10 to 1000ÎĽL), 1 x Linear Stand and 4 x Pipet Holders
Multipacks combine a set of Proline Plus single-channel mechanical pipettes with matching tip trays and useful accessories. Sartorius Proline Pus Mechanical Pipette Multipacks provides best alternative to replace your used mechanical pipettes.
  • Low pipetting forces
  • Ergonomic finger hook
  • Easy volume setting with click stop mechanism
  • Quick and easy-to-read volume display minimises risk of volume setting errors
  • Thermal insulation of internal components improves accuracy
  • Easy to use 'do-it-yourself' calibration
  • Easy to clean with only three parts to disassemble
  • Fully autoclavable
  • High chemical and UV-resistance
  • High-quality standard Optifit tips for various applications
  • Optiload tip loading mechanism allows tip loading with an equal constant force onto every channel and enables perfect tip sealing onto every individual tip cone
Warranty
Two years
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Optifit Tips
Autoclavable Autoclavable
Disposable Reusable
Model Proline Plus
Number of Channels 1
Product Line Proline Plus
Sterility Non-sterile
Type Mechanical Pipette Multipacks
Volume (Metric) 0.5 to 10 ÎĽL, 2 to 20 ÎĽL, 20 to 200 ÎĽL, 100 to 1000 ÎĽL
Filter Type Safe-Cone Filter
Includes 4 x Single-channel pipets (0.5 to 10ÎĽL, 2 to 20ÎĽL, 20 to 200ÎĽL, 100 to 1000ÎĽL), Tip Racks of 96 tips (0.1 to 10ÎĽL, 0.5 to 200ÎĽL, 10 to 1000ÎĽL), 1 x Linear Stand and 4 x Pipet Holders
Quantity 10/Pk.
UV Monitoring UV resistant
Warranty 2 Years
Product Title
Sartorius Proline Plus Mechanical Pipette Multipacks >