Learn More
Ohaus™ OIML E1 Weight Set
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand:  Ohaus™ 30517993
Additional Details : Weight : 6.11000kg
 View more versions of this product
Product Code. 16642332
  • 65975.00 NOK / Each
Estimated Shipment: 21-06-2024
Description
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
Specifications
No Certificate
2 x 2 kg to 1 mg
27/Pk.
Troemner Alloy 8, Stainless Steel, Aluminum
Protective Case