Learn More
Ohaus™ OIML E1 Weight Set
Adjusted to Meet or Exceed OIML E1 Tolerances
12205.00 NOK - 107880.00 NOK
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16682322
|
Ohaus™
30517987 |
500 mg to 1 mg |
12205.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16692322
|
Ohaus™
30517988 |
50 g to 1 mg |
24295.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16672332
|
Ohaus™
30517996 |
50 g to 1 g |
14510.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16602332
|
Ohaus™
30517989 |
100 g to 1 mg |
26715.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16682332
|
Ohaus™
30517997 |
100 g to 1 g |
16925.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16612332
|
Ohaus™
30517990 |
2 x 200 g to 1 mg |
33045.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16692332
|
Ohaus™
30517998 |
2 x 200 g to 1 g |
23030.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16622332
|
Ohaus™
30517991 |
500 g to 1 mg |
38225.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16602342
|
Ohaus™
30517999 |
500 g to 1 g |
28325.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16632332
|
Ohaus™
30517992 |
1 kg to 1 mg |
45135.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16612342
|
Ohaus™
30518000 |
1 kg to 1 g |
34885.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16642332
|
Ohaus™
30517993 |
2 x 2 kg to 1 mg |
65975.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16622342
|
Ohaus™
30518001 |
2 x 2 kg to 1 g |
55035.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16652332
|
Ohaus™
30517994 |
5 kg to 1 mg |
83360.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16632342
|
Ohaus™
30518002 |
5 kg to 1 g |
73110.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16662332
|
Ohaus™
30517995 |
10 kg to 1 mg |
107880.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
16642342
|
Ohaus™
30518003 |
10 kg to 1 g |
97980.00 NOK
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
Protective Case |